Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triisobutylaluminium, 1.0M solution in toluene, AcroSeal™

Catalog No. AC185428000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC185428000 800 mL AcroSeal™ Glass Bottle
AC185421000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC185428000 Supplier Thermo Scientific Chemicals Supplier No. 185428000
Only null left
Add to Cart

CAS: 100-99-2 | C12H27Al | 198.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-99-2 , 108-88-3
Molecular Formula C12H27Al
Molecular Weight (g/mol) 198.33
MDL Number MFCD00008929
InChI Key MCULRUJILOGHCJ-UHFFFAOYSA-N
Synonym triisobutylaluminum, triisobutylaluminium, aluminum, tris 2-methylpropyl, tibal, triisobutylalane, triisobutyl aluminum, aluminum, triisobutyl, tris isobutyl alane, tris isobutyl aluminum, unii-09p2thv2x4
PubChem CID 16682931
IUPAC Name tris(2-methylpropyl)alumane
SMILES CC(C)C[Al](CC(C)C)CC(C)C

Specifications

Color Colorless to Yellow
Linear Formula [(CH3)2CHCH2]3Al
Fieser 09,486
Density 0.8400g/mL
Merck Index 10, 316
Solubility Information Solubility in water: reacts. Other solubilities: miscible with aromatic and aliphatic hydrocarbons
Formula Weight 198.33
Percent Purity 24.5 to 25.5% (on Al basis)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.84
Quantity 800 mL
Flash Point 4°C
Physical Form Solution
Chemical Name or Material Triisobutylaluminium, 1.1M Solution in Toluene
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.