Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D386125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3861-25G 25 g
D38615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3861-25G Supplier TCI America Supplier No. D386125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23112-96-1
Molecular Formula C8H11BO4
MDL Number MFCD01318987
InChI Key BKWVXPCYDRURMK-UHFFFAOYSA-N
PubChem CID 2734343
IUPAC Name (2,6-dimethoxyphenyl)boronic acid

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 25 g
Synonym 2,6-dimethoxyphenyl boronic acid, 2,6-dimethoxyphenylboronicacid, 2,6-dimethoxybenzeneboronic acid, 2,6-dimethoxyphenyl-boronic acid, 2,6-dimethoxyphenyl boranediol
SMILES COC1=CC=CC(OC)=C1B(O)O
Molecular Weight (g/mol) 181.98
Formula Weight 181.98
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.