Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxypyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D46905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4690-5G 5 g
D46901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4690-5G Supplier TCI America Supplier No. D46905G
Add to Cart
Add to Cart

Specifications

CAS 221006-70-8
Melting Point 109°C
Color White
Molecular Formula C7H10BNO4
Quantity 5 g
MDL Number MFCD03788239
Synonym 2,6-dimethoxypyridine-3-boronic acid, 2,6-dimethoxy-3-pyridineboronic acid, 2,6-dimethoxypyridin-3-yl boronic acid, 2,6-dimethoxypyridin-3-yl-3-boronic acid, 3-borono-2,6-dimethoxypyridine
InChI Key ADGHSWFUZUADDH-UHFFFAOYSA-N
SMILES B(C1=C(N=C(C=C1)OC)OC)(O)O
IUPAC Name (2,6-dimethoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 182.97
PubChem CID 2762707
Formula Weight 182.97
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.