Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxypyridine-3-boronic acid pinacol ester, 97%

Catalog No. AC444920050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC444920050 5 g
AC444920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444920050 Supplier Thermo Scientific Chemicals Supplier No. 444920050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 214360-59-5 | C13H20BNO4 | 265.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214360-59-5
Molecular Formula C13H20BNO4
Molecular Weight (g/mol) 265.12
MDL Number MFCD03788240
InChI Key VZGDCNFVMWRPOW-UHFFFAOYSA-N
Synonym 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxypyridine-3-boronic acid pinacol ester, 2,6-dimethoxypyridine-5-boronic acid pinacol ester, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2,6-dimethoxypyridine-3-boronic acid, pinacol ester, 2,6-dimethoxy-5 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, pyridine, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2,6-dimethoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2,6-dimethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxabo
PubChem CID 2758487
IUPAC Name 2,6-dimethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES COC1=CC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1

Specifications

Melting Point < 39°C to 44°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 265.12
Percent Purity 97%
Chemical Name or Material 2, 6-Dimethoxypyridine-3-boronic acid pinacol ester
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.