Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethoxypyridine 97.0+%, TCI America™
SDP

Catalog No. D191825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D191825G Supplier TCI America Supplier No. D191825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6231-18-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.15
MDL Number MFCD00006266
InChI Key IBTGEEMBZJBBSH-UHFFFAOYSA-N
Synonym pyridine, 2,6-dimethoxy, 2,6-dimethoxy pyridine, pyridine,2,6-dimethoxy, acmc-1b7tt, ksc494c3n, 2,6-dimethoxypyridine, 2,6-dimethoxypyridine 25g
PubChem CID 80378
IUPAC Name 2,6-dimethoxypyridine
SMILES COC1=CC=CC(OC)=N1

Specifications

Color Colorless
Boiling Point 179°C
Quantity 25 g
Formula Weight 139.15
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Dimethoxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.