Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dimethyl-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. N099725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N099725G 25 g
N09975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N099725G Supplier TCI America Supplier No. N099725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15513-52-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00023503
InChI Key AETHUDGJSSKZKT-UHFFFAOYSA-N
PubChem CID 84958
IUPAC Name 2,6-dimethyl-3-nitropyridine
SMILES CC1=NC(=C(C=C1)[N+](=O)[O-])C

Specifications

Melting Point 38°C
Color Yellow
Boiling Point 105°C
Quantity 25 g
UN Number 1325
Synonym 3-nitro-2,6-lutidine, 3-nitro-2,6-dimethylpyridine, pyridine, 2,6-dimethyl-3-nitro, nitrolutidine, pubchem14242, acmc-20a0s3, 2,6-dimethyl-3-nitro-pyridine, pyridine,2,6-dimethyl-3-nitro, 2-methyl-5-nitro-6-methylpyridine
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethyl-3-nitropyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.