Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,6-Dimethylphenyl)thiourea 98.0+%, TCI America™
SDP

Catalog No. D495825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D495825G 25 g
D49585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D495825G Supplier TCI America Supplier No. D495825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6396-76-5
Molecular Formula C9H12N2S
Molecular Weight (g/mol) 180.27
MDL Number MFCD00041165
InChI Key ASNKJUONFPQYPC-UHFFFAOYSA-N
Synonym 1-2,6-dimethylphenyl thiourea, 1-2,6-dimethylphenyl-2-thiourea, 2,6-dimethylphenyl thiourea, n-2,6-dimethylphenyl thiourea, 1-2,6-xylyl thiourea, thiourea, 2,6-dimethylphenyl, amino 2,6-dimethylphenyl amino methane-1-thione, acmc-20apd5, 1-2,6-xylyl-2-thiourea, 2,6-dimethyl-phenyl-thiourea
PubChem CID 853911
IUPAC Name (2,6-dimethylphenyl)thiourea
SMILES CC1=CC=CC(C)=C1NC(N)=S

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 180.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (2,6-Dimethylphenyl)thiourea
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.