Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diphenylphenol, 98%

Catalog No. AAL1716606
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1716606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1716606 Supplier Thermo Scientific Chemicals Supplier No. L1716606

missing translation for 'validMainframeMsg'

CAS: 2432-11-3 | C5H2Br2O3 | 269.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2432-11-3
MDL Number MFCD00009716

Specifications

Melting Point 101°C to 103°C
Molecular Formula C18H14O
Quantity 5 g
Beilstein 2051224
Synonym 1,1':3',1-terphenyl-2'-ol, m-terphenyl-2'-ol, 2,6-diphenyl phenol, 2'-hydroxy-m-terphenyl, unii-la93w35m96, 3-phenyl-1,1'-biphenyl-2-ol, 2,6-di-phenylphenol, 2,6,-diphenylphenol, 2,6-di phenyl phenol
InChI Key ATGFTMUSEPZNJD-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=C(C(=CC=C2)C3=CC=CC=C3)O
IUPAC Name 2,6-diphenylphenol
Molecular Weight (g/mol) 246.31
PubChem CID 75512
Formula Weight 246.31
Percent Purity 98%
Chemical Name or Material 2,6-Diphenylphenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-401-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.