Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diphenylpyridine, 97%, Thermo Scientific™

Catalog No. AC189670100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC189670100 10 g
AC189670500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC189670100 Supplier Thermo Scientific Chemicals Supplier No. 189670100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3558-69-8
Molecular Formula C17H13N
Molecular Weight (g/mol) 231.298
InChI Key PJUOHDQXFNPPRF-UHFFFAOYSA-N
Synonym pyridine, 2,6-diphenyl, unii-zi75v15y1t, 2,6-diphenyl-pyridine, 2,6-diphenyl pyridine, maybridge3_000511, acmc-1cq41, pjuohdqxfnpprf-uhfffaoysa
PubChem CID 72920
IUPAC Name 2,6-diphenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)C3=CC=CC=C3

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 73°C to 77°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula C6H5(C5H3N)C6H5
Quantity 10 g
Beilstein 20, 496
Formula Weight 231.3
Percent Purity ≥96.0%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 6-Diphenylpyridine, 97%
Show More Show Less
EINECSNumber 222-620-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.