Learn More
Sigma Organic Chemistry 2- 6-Fluoro-1H-benzimid | 1G | MFCD08059825
Supplier: Sigma Organic Chemistry CBR010951G

2- 6-Fluoro-1H-benzimid , 1G
About this Item:
MDL #: MFCD08059825
Molecular Weight: 180.18
UNSPSC Code: 12352200
Chemical Formula: C9H9FN2O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.