Learn More
Sigma Organic Chemistry 2,6-Lutidine | 500ML | 108-48-5 | MFCD00006345
Supplier: Sigma Organic Chemistry L3900500MLA

2,6-Lutidine, 500ML
About This Item:
Synonym(s): 2,6-Dimethylpyridine
Linear Formula: C7H9N
Molecular Weight: 107.15
CAS Number: 108-48-5
MDL Number: MFCD00006345
UNSPSC Code: 12352005
Purity: 0.98
Boiling Point: 143-145 C (lit.)
Melting Point: -6 C (lit.)
Density: 0.92 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.497 (lit.)
Storage: room temp
EINECS Number: 203-587-3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.