Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Pyridinedicarboxylic acid chloride, 97%

Catalog No. AC162130250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC162130250 25 g Glass bottle
AC162130050 5 g Glass bottle
AC162131000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC162130250 Supplier Thermo Scientific Chemicals Supplier No. 162130250
Add to Cart

CAS: 3739-94-4 | C7H3Cl2NO2 | 204.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3739-94-4
Molecular Formula C7H3Cl2NO2
Molecular Weight (g/mol) 204.01
MDL Number MFCD00006289
InChI Key GWHOGODUVLQCEB-UHFFFAOYSA-N
Synonym 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153
PubChem CID 77335
IUPAC Name pyridine-2,6-dicarbonyl chloride
SMILES C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 57.0°C to 61.0°C
Color White to Brown
Boiling Point 284.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H3N)(COCl)2
Quantity 25 g
Beilstein 22, 155
Formula Weight 204.01
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2, 6-Pyridinedicarboxylic acid chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 223-125-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.