Learn More
AdipoGen 2 6-TNS
SDP
Supplier:  AdipoGen CDXT0008M250
Encompass_Preferred
Chemical. CAS 7724-15-4. Formula C17H15NO3S. Molecular Weight 313.37. 6-p-Toluidino-2-naphthalenesulfonic acid TNS is a widely recognized aminonaphthalene-based fluorescent probe characterized for its solvatochromic effect. TNS is found to be almost fluorescence silent in aqueous medium whereas shows a high quantum yield in organic solvents with a hypsochromic shift, as the polarity of the medium decreases. This property has been exploited in biological studies to probe the micellation properties of surfactants, membrane fluidity, hydrophobic surfaces on proteins, conformational changes upon ligand binding, ligand binding events, protein-surfactant interactions, multimeric protein assembly or aggregation, fibril formation and others. TNS has been used in many studies as environmentally sensitive fluorescent probe for the conformational state of proteins.
Catalog No. 50-256-0050
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.