Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Bis[N-(1-naphthyl)anilino]-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
SDP

Catalog No. B48751G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48751G 1 g
B4875200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48751G Supplier TCI America Supplier No. B48751G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 932739-76-9
Molecular Formula C57H38N2
Molecular Weight (g/mol) 750.945
MDL Number MFCD12022428
InChI Key ZDAWFMCVTXSZTC-UHFFFAOYSA-N
PubChem CID 51358306
IUPAC Name 2-N',7-N'-dinaphthalen-1-yl-2-N',7-N'-diphenyl-9,9'-spirobi[fluorene]-2',7'-diamine
SMILES C1=CC=C(C=C1)N(C2=CC3=C(C=C2)C4=C(C35C6=CC=CC=C6C7=CC=CC=C57)C=C(C=C4)N(C8=CC=CC=C8)C9=CC=CC1=CC=CC=C19)C1=CC=CC2=CC=CC=C21

Specifications

Melting Point 284°C
Color White-Yellow
Quantity 1 g
Formula Weight 750.95
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis[N-(1-naphthyl)anilino]-9,9′-spirobi[9H-fluorene]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.