Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Di-tert-butylfluorene 98.0+%, TCI America™
SDP

Catalog No. D408425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D408425G 25 g
D40845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D408425G Supplier TCI America Supplier No. D408425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58775-05-6
Molecular Formula C21H26
Molecular Weight (g/mol) 278.439
MDL Number MFCD03093998
InChI Key DFZYPLLGAQIQTD-UHFFFAOYSA-N
Synonym 2,7-di-tert-butylfluorene, 2,7-di-tert-butyl-9h-fluorene, 2,7-di-t-butylfluorene, 9h-fluorene, 2,7-bis 1,1-dimethylethyl, pubchem15066, 2,7-di-tert-butyl fluorene, 2,7-di-tert-butyl-fluorene, ksc491i2b
PubChem CID 4090082
IUPAC Name 2,7-ditert-butyl-9H-fluorene
SMILES CC(C)(C)C1=CC2=C(C=C1)C3=C(C2)C=C(C=C3)C(C)(C)C

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 25 g
Formula Weight 278.44
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Di-tert-butylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.