Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. D52701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D52701G 1 g
D5270200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D52701G Supplier TCI America Supplier No. D52701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1313900-20-7
Molecular Formula C18H10Br3N
Molecular Weight (g/mol) 479.997
InChI Key MKSHSKZVLIHXMT-UHFFFAOYSA-N
PubChem CID 129317820
IUPAC Name 2,7-dibromo-9-(4-bromophenyl)carbazole
SMILES C1=CC(=CC=C1N2C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br)Br

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 1 g
Formula Weight 480.00
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-(4-bromophenyl)-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.