Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9,9-di-n-octylfluorene 98.0+%, TCI America™
SDP

Catalog No. D393425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D393425G 25 g
D39345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D393425G Supplier TCI America Supplier No. D393425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 198964-46-4
Molecular Formula C29H40Br2
Molecular Weight (g/mol) 548.45
MDL Number MFCD03427216
InChI Key CYKLQIOPIMZZBZ-UHFFFAOYSA-N
PubChem CID 5215321
IUPAC Name 2,7-dibromo-9,9-dioctyl-9H-fluorene
SMILES CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 54°C
Color White-Yellow
Quantity 25 g
Formula Weight 548.45
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,7-Dibromo-9,9-di-n-octylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.