Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9,9-didecylfluorene 98.0+%, TCI America™
SDP

Catalog No. D456125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D456125G 25 g
D45615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D456125G Supplier TCI America Supplier No. D456125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175922-78-8
Molecular Formula C33H48Br2
Molecular Weight (g/mol) 604.555
MDL Number MFCD00971288
InChI Key RLYANTSRYXOALQ-UHFFFAOYSA-N
PubChem CID 3661211
IUPAC Name 2,7-dibromo-9,9-didecylfluorene
SMILES CCCCCCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCCCCCC

Specifications

Melting Point 40°C
Color White-Yellow
Quantity 25 g
Formula Weight 604.56
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-didecylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.