Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9,9-diphenylfluorene 98.0+%, TCI America™
SDP

Catalog No. D45471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45471G 1 g
D45475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D45471G Supplier TCI America Supplier No. D45471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 186259-63-2
Molecular Formula C25H16Br2
Molecular Weight (g/mol) 476.211
MDL Number MFCD09907958
InChI Key AJYDOCCGNIBJBY-UHFFFAOYSA-N
PubChem CID 16744636
IUPAC Name 2,7-dibromo-9,9-diphenylfluorene
SMILES C1=CC=C(C=C1)C2(C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br)C5=CC=CC=C5

Specifications

Melting Point 279°C
Quantity 1 g
Formula Weight 476.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9,9-diphenylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.