Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9-fluorenone 98.0+%, TCI America™
SDP

Catalog No. D355725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D355725G 25 g
D35575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D355725G Supplier TCI America Supplier No. D355725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14348-75-5
Molecular Formula C13H6Br2O
Molecular Weight (g/mol) 337.998
MDL Number MFCD00010790
InChI Key CWGRCRZFJOXQFV-UHFFFAOYSA-N
Synonym 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427
PubChem CID 259922
IUPAC Name 2,7-dibromofluoren-9-one
SMILES C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br

Specifications

Melting Point 204°C
Color Yellow
Quantity 25 g
Formula Weight 338.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-fluorenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.