Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9-heptyl-9H-carbazole 98.0+%, TCI America™
SDP

Catalog No. D54031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54031G 1 g
D54035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54031G Supplier TCI America Supplier No. D54031G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1173071-58-3
Molecular Formula C19H21Br2N
Molecular Weight (g/mol) 423.192
InChI Key MLLYLIXYBBORGG-UHFFFAOYSA-N
PubChem CID 129319314
IUPAC Name 2,7-dibromo-9-heptylcarbazole
SMILES CCCCCCCN1C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br

Specifications

Melting Point 74°C
Color White
Quantity 1 g
Formula Weight 423.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-heptyl-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.