Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',7'-Dichlorofluorescein 95.0+%, TCI America™
SDP

Catalog No. D03711G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D03711G 1 g
D037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D03711G Supplier TCI America Supplier No. D03711G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 76-54-0
Molecular Formula C20H10Cl2O5
Molecular Weight (g/mol) 401.195
MDL Number MFCD00005047
InChI Key VFNKZQNIXUFLBC-UHFFFAOYSA-N
Synonym 2',7'-dichlorofluorescein, dichlorofluorescein, unii-56nqm5uzt1, 2,7-dichlorofluorescein, 56nqm5uzt1, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxy, 2',7'-dichloro-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 2',7'-dichloro-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 11,16-dichloro-12,15-dihydroxyspiro 3-hydroisobenzofuran-3,9'-xanthene-1-one
PubChem CID 64944
ChEBI CHEBI:51596
IUPAC Name 2',7'-dichloro-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C=C4OC5=CC(=C(C=C35)Cl)O)O)Cl

Specifications

Melting Point 280°C
Quantity 1 g
Formula Weight 401.20
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2′,7′-Dichlorofluorescein [for Fluorescent indicator]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.