Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dimethoxynaphthalene 98.0+%, TCI America™
SDP

Catalog No. D191725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D191725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D191725G Supplier TCI America Supplier No. D191725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3469-26-9
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00004064
InChI Key PPKHAIRFQKFMLE-UHFFFAOYSA-N
Synonym naphthalene, 2,7-dimethoxy, acmc-209i9n, ksc225e3r, 2,7-dimethoxynaphthalene, ppkhairfqkfmle-uhfffaoysa, 2,7-dimethoxy naphthalene, benzocyclobuten-1 2h-one; bicyclo 4.2.0 octa-1,3,5-trien-7-one
PubChem CID 77021
IUPAC Name 2,7-dimethoxynaphthalene
SMILES COC1=CC2=C(C=C1)C=CC(=C2)OC

Specifications

Melting Point 139°C
Color White-Yellow
Quantity 25 g
Formula Weight 188.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dimethoxynaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.