Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,8-Dibromodibenzothiophene 5,5-Dioxide 95.0+%, TCI America™
SDP

Catalog No. D50271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50271G 1 g
D5027200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50271G Supplier TCI America Supplier No. D50271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40307-15-1
Molecular Formula C12H6Br2O2S
Molecular Weight (g/mol) 374.046
MDL Number MFCD00093102
InChI Key ZFGCKZCEDNBNMV-UHFFFAOYSA-N
Synonym 2,8-Dibromodibenzothiophene Sulfone, 2,8-Dibromodibenzosulfolane
PubChem CID 3905994
IUPAC Name 2,8-dibromodibenzothiophene 5,5-dioxide
SMILES C1=CC2=C(C=C1Br)C3=C(S2(=O)=O)C=CC(=C3)Br

Specifications

Melting Point 361°C
Quantity 1 g
Formula Weight 374.05
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dibromodibenzothiophene 5,5-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.