Learn More
Azzota Corp 2C TO 8C PHARMACY & MEDICAL LAB REFRIGERATOR, UL CERTIFICATION, 110V/60HZ, 650L, internal 605*725*1515mm
Supplier:  Azzota Corp YC650CL
Encompass_Preferred
2C TO 8C PHARMACY & MEDICAL LAB REFRIGERATOR, UL CERTIFICATION, 110V/60HZ, 650L, internal 605*725*1515mm
Catalog No. NC2753692
Only null left
Add to Cart
Request a Quote