Learn More
Sigma Organic Chemistry 2- acetoacetyl phenol | 100mg | 16636-62-7 | MFCD00013967
Supplier: Sigma Organic Chemistry CDS008776100MG

2- acetoacetyl phenol , 100mg
About this Item:
CAS #: 16636-62-7
MDL #: MFCD00013967
Molecular Weight: 178.18
UNSPSC Code: 12352200
Chemical Formula: C10H10O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.