Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Acetonaphthone 98.0+%, TCI America™
SDP

Catalog No. A0053100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0053100G 100 g
A005325G 25 g
A0053500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0053100G Supplier TCI America Supplier No. A0053100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93-08-3
Molecular Formula C12H10O
Molecular Weight (g/mol) 170.211
MDL Number MFCD00004108
InChI Key XSAYZAUNJMRRIR-UHFFFAOYSA-N
Synonym 2-acetylnaphthalene, 2-acetonaphthone, 2'-acetonaphthone, methyl 2-naphthyl ketone, acetonaphthone, 1-naphthalen-2-yl ethanone, 1-2-naphthyl ethanone, ethanone, 1-2-naphthalenyl, oranger cyrstals, 2-naphthyl methyl ketone
PubChem CID 7122
ChEBI CHEBI:52364
IUPAC Name 1-naphthalen-2-ylethanone
SMILES CC(=O)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 301°C
Quantity 100 g
Formula Weight 170.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Acetonaphthone
EINECSNumber (4)-0394
RTECSNumber AL2988000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.