Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Acetoxyacetophenone, 98%

Catalog No. AAB2162414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2162414 25 g
AAB2162406 5 g
AAB2162422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2162414 Supplier Thermo Scientific Chemicals Supplier No. B2162414
Add to Cart

CAS: 7250-94-4 | C10H10O3 | 178.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7250-94-4
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017223
InChI Key WWEDAIJJBDFOOK-UHFFFAOYSA-N
Synonym 2'-acetoxyacetophenone, 2-acetylphenyl acetate, ethanone, 1-2-acetyloxy phenyl, o-acetoxyacetophenone, 2'-acetoxy acetophenone, maybridge1_002653, acmc-209on8, acetic acid 2-acetyl-phenyl ester, acetophenone, 2-hydroxy-, acetate 8ci
PubChem CID 16716
IUPAC Name (2-acetylphenyl) acetate
SMILES CC(=O)OC1=CC=CC=C1C(C)=O

Specifications

Melting Point 88°C to 91°C
Boiling Point 118°C (19 mmHg)
Quantity 25 g
Beilstein 1948938
Formula Weight 178.19
Percent Purity 98%
Chemical Name or Material 2'-Acetoxyacetophenone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.