Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-3-bromothiophene, 97%, Thermo Scientific™

Catalog No. AAL2011303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2011303 1 g
AAL2011306 5 g
AAL2011314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL2011303 Supplier Thermo Scientific Chemicals Supplier No. L2011303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42877-08-7
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.07
MDL Number MFCD00794287
InChI Key ZUXPELAHJQSZTE-UHFFFAOYSA-N
Synonym 2-acetyl-3-bromothiophene, 1-3-bromothiophen-2-yl ethanone, 1-3-bromothiophen-2-yl ethan-1-one, 1-3-bromo-2-thienyl-1-ethanone, bromothienylethanone, 1-3-bromothien-2-yl ethanone, 1-3-bromo-2-thienyl ethanone, pubchem7280, acmc-20ai7w
PubChem CID 2764496
SMILES CC(=O)C1=C(Br)C=CS1

Specifications

Melting Point 41°C to 45°C
Boiling Point 102°C to 105°C (4 mmHg)
Refractive Index 1.614
Quantity 1 g
Beilstein 116973
Sensitivity Light sensitive
IUPAC Name 1-(3-bromothiophen-2-yl)ethanone
Formula Weight 205.07
Percent Purity 97%
Chemical Name or Material 2-Acetyl-3-bromothiophene
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.