Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione 98.0+%, TCI America™
SDP

Catalog No. A18531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18531G 1 g
A18535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18531G Supplier TCI America Supplier No. A18531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1755-15-3
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00467663
InChI Key ITSKWKZDPHAQNK-UHFFFAOYSA-N
Synonym 2-Acetyldimedone
PubChem CID 519448
IUPAC Name 2-acetyl-5,5-dimethylcyclohexane-1,3-dione
SMILES CC(=O)C1C(=O)CC(C)(C)CC1=O

Specifications

Melting Point 36°C
Color Yellow
Quantity 1 g
Formula Weight 182.22
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Acetyl-5,5-dimethyl-1,3-cyclohexanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.