Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-5-bromothiophene 98.0+%, TCI America™
SDP

Catalog No. A118325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A118325G 25 g
A11835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A118325G Supplier TCI America Supplier No. A118325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5370-25-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.069
MDL Number MFCD00014528
InChI Key IGBZCOWXSCWSHO-UHFFFAOYSA-N
Synonym 2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
PubChem CID 79335
IUPAC Name 1-(5-bromothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Br

Specifications

Melting Point 95°C
Boiling Point 103°C
Quantity 25 g
Formula Weight 205.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-5-bromothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.