Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-6-methoxynaphthalene 98.0+%, TCI America™
SDP

Catalog No. A2030100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2030100G 100 g
A203025G 25 g
A2030500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A2030100G Supplier TCI America Supplier No. A2030100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3900-45-6
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00021643
InChI Key GGWCZBGAIGGTDA-UHFFFAOYSA-N
Synonym 2-acetyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethanone, 1-6-methoxy-2-naphthyl ethanone, 6-methoxy-2-acetylnaphthalene, unii-61vbh3537c, 6'-methoxy-2'-acetonaphthone, 1-6-methoxy-2-naphthyl ethan-1-one, ethanone, 1-6-methoxy-2-naphthalenyl, 2-acetyl-6-methoxy naphthalene, 6-methoxy-2-acetyl naphthalene
PubChem CID 77506
IUPAC Name 1-(6-methoxynaphthalen-2-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)C=C(C=C2)OC

Specifications

Melting Point 109°C
Color Yellow
Boiling Point 195°C
Quantity 100 g
Formula Weight 200.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-6-methoxynaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.