Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetyl-7-hydroxybenzofuran 98.0+%, TCI America™
SDP

Catalog No. A131610G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A131610G 10 g
A13165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A131610G Supplier TCI America Supplier No. A131610G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40020-87-9
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00191373
InChI Key HRIVIHCUSKDPNK-UHFFFAOYSA-N
Synonym 2-Acetyl-7-hydroxycoumarone
PubChem CID 595329
IUPAC Name 1-(7-hydroxy-1-benzofuran-2-yl)ethan-1-one
SMILES CC(=O)C1=CC2=CC=CC(O)=C2O1

Specifications

Melting Point 166°C
Color Yellow
Quantity 10 g
Formula Weight 176.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-7-hydroxybenzofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.