Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylbenzo[b]thiophene 98.0+%, TCI America™
SDP

Catalog No. A149725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A149725G 25 g
A14975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A149725G Supplier TCI America Supplier No. A149725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22720-75-8
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.23
MDL Number MFCD00090217
InChI Key SGSGCQGCVKWRNM-UHFFFAOYSA-N
Synonym 2-acetylbenzothiophene, 2-acetylbenzo b thiophene, 1-benzo b thiophen-2-yl ethanone, 1-benzo b thiophen-2-yl-ethanone, 1-1-benzothiophen-2-yl ethanone, 1-1-benzothiophen-2-yl ethan-1-one, ethanone, 1-benzo b thienyl, 1-benzo b thiophen-2-yl ethan-1-one
PubChem CID 89805
IUPAC Name 1-(1-benzothiophen-2-yl)ethan-1-one
SMILES CC(=O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point 89°C
Quantity 25 g
UN Number 3077
Formula Weight 176.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylbenzo[b]thiophene
EINECSNumber (5)-6262
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.