Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylpyrrole 98.0+%, TCI America™
SDP

Catalog No. A089425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A089425G 25 g
A08945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A089425G Supplier TCI America Supplier No. A089425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1072-83-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00005220
InChI Key IGJQUJNPMOYEJY-UHFFFAOYSA-N
Synonym 2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl
PubChem CID 14079
ChEBI CHEBI:59981
IUPAC Name 1-(1H-pyrrol-2-yl)ethanone
SMILES CC(=O)C1=CC=CN1

Specifications

Melting Point 90°C
Color White
Boiling Point 220°C
Quantity 25 g
Formula Weight 109.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylpyrrole
EINECSNumber (5)-0101
RTECSNumber OB5970000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.