Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Adamantanone Oxime 98.0+%, TCI America™
SDP

Catalog No. A211025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A211025G 25 g
A21105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A211025G Supplier TCI America Supplier No. A211025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4500-12-3
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD00078273
InChI Key RABVIFXMFZFITE-KHPPLWFESA-N
Synonym 2-adamantanone oxime, adamantan-2-one oxime, adamantanone oxime, 2-adamantanone, oxime, 2-hydroxyiminoadamantane, adamantanone-2-oxime, 2-isonitrosoadamantane, 2-hydroxyimino adamantane, n-2-adamantylidene hydroxylamine, n-adamantan-2-ylidene hydroxylamine
PubChem CID 64158
IUPAC Name N-(2-adamantylidene)hydroxylamine
SMILES C1C2CC3CC1CC(C2)C3=NO

Specifications

Melting Point 168°C
Color White
Quantity 25 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Adamantanone Oxime
RTECSNumber AU5018500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.