Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Amidoximophenyl)acetamidoxime, 97%, Thermo Scientific™

Catalog No. AAH5897404
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5897404 2 g
AAH5897409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5897404 Supplier Thermo Scientific Chemicals Supplier No. H5897404

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1421266-64-9
Molecular Formula C9H12N4O2
Molecular Weight (g/mol) 208.221
MDL Number MFCD22988982
InChI Key CTCKPOCZAHVUEJ-UHFFFAOYSA-N
Synonym 2-amidoximophenyl acetamidoxime
PubChem CID 121235634
IUPAC Name 2-[(2E)-2-amino-2-hydroxyiminoethyl]-N'-hydroxybenzenecarboximidamide
SMILES C1=CC=C(C(=C1)CC(=NO)N)C(=NO)N

Specifications

Quantity 2 g
Formula Weight 208.21
Percent Purity 97%
Chemical Name or Material 2-(Amidoximophenyl)acetamidoxime
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.