Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-1,3-propanediol 98.0+%, TCI America™
SDP

Catalog No. A125225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A125225G 25 g
A12525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A125225G Supplier TCI America Supplier No. A125225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 534-03-2
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00051487
InChI Key KJJPLEZQSCZCKE-UHFFFAOYSA-N
Synonym 2-amino-1,3-propanediol, serinol, 1,3-propanediol, 2-amino, 2-amino-1,3-propandiol, 2-aminopropan-1,3-diol, unii-ic94l30j8m, 2-amino-1,3-dihydroxypropane, 2-amino-1,3-propanediol serinol, dl-serinol, sel
PubChem CID 68294
IUPAC Name 2-aminopropane-1,3-diol
SMILES NC(CO)CO

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 116°C
Quantity 25 g
UN Number 3259
Formula Weight 91.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-1,3-propanediol
EINECSNumber (2)-3541
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.