Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2-Amino-1-phenylethanol, 95%

Catalog No. ac104320100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104320100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC104320100 Supplier Thermo Scientific Chemicals Supplier No. 104320100
Only null left
Add to Cart

CAS: 7568-93-6 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7568-93-6
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008137
InChI Key ULSIYEODSMZIPX-UHFFFAOYNA-N
Synonym phenylethanolamine, 2-hydroxy-2-phenylethylamine, phenethanolamine, beta-hydroxyphenethylamine, 2-phenyl-2-hydroxyethylamine, bisnorephedrine, beta-phenethanolamine, alpha-aminomethyl benzyl alcohol, 2-amino-1-phenylethan-1-ol, 2-amino-1-phenyl-1-ethanol
PubChem CID 1000
ChEBI CHEBI:16343
IUPAC Name 2-amino-1-phenylethanol
SMILES NCC(O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color Yellow
Boiling Point 160.0°C (17.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.5580 to 1.5600
Linear Formula H2NCH2CH(C6H5)OH
Quantity 10 g
Merck Index 15, 7399
Formula Weight 137.18
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material (±)-2-Amino-1-phenylethanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 231-469-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.