Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2'-chloro-5-nitrobenzophenone 98.0+%, TCI America™
SDP

Catalog No. A175425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A175425G 25 g
A17545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A175425G Supplier TCI America Supplier No. A175425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2011-66-7
Molecular Formula C13H9ClN2O3
Molecular Weight (g/mol) 276.68
MDL Number MFCD00792453
InChI Key GRDGBWVSVMLKBV-UHFFFAOYSA-N
Synonym 2-amino-2'-chloro-5-nitrobenzophenone, 2-amino-5-nitrophenyl 2-chlorophenyl methanone, 2-amino-5-nitro-2'-chlorobenzophenone, 2-amino-2'-chloro-5-nitro benzophenone, 2-2-chlorobenzoyl-4-nitroaniline, methanone, 2-amino-5-nitrophenyl 2-chlorophenyl, 2-amino-5-nitrophenyl-2-chlorophenyl methanone, 2-amino-5-nitro-phenyl-2-chloro-phenyl-methanone, 2-amino-5-nitro-2'-chloro benzophenone, pubchem3255
PubChem CID 74830
IUPAC Name 2-(2-chlorobenzoyl)-4-nitroaniline
SMILES NC1=CC=C(C=C1C(=O)C1=CC=CC=C1Cl)[N+]([O-])=O

Specifications

Melting Point 121°C
Color Yellow
Quantity 25 g
Formula Weight 276.68
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2′-chloro-5-nitrobenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.