Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-2'-nitrodiphenyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. A198525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A198525G 25 g
A19855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A198525G Supplier TCI America Supplier No. A198525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19284-81-2
Molecular Formula C12H10N2O2S
Molecular Weight (g/mol) 246.28
MDL Number MFCD00121816
InChI Key QREMWHOIUTWWSC-UHFFFAOYSA-N
Synonym 2-(2-Nitrophenylthio)aniline, 2-Aminophenyl 2-Nitrophenyl Sulfide
PubChem CID 699904
IUPAC Name 2-[(2-nitrophenyl)sulfanyl]aniline
SMILES NC1=CC=CC=C1SC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 87°C
Color Yellow
Quantity 25 g
Formula Weight 246.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-2′-nitrodiphenyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.