Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,4-dimethylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. D412025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D412025G 25 g
D41205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D412025G Supplier TCI America Supplier No. D412025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50419-58-4
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00130041
InChI Key MUOBMUYSNYMSDM-UHFFFAOYSA-N
Synonym 2-amino-3,4-dimethyl-benzoic acid, 3,4-dimethylanthranilic acid, benzoic acid, 2-amino-3,4-dimethyl, pubchem4683, acmc-1aomr, ksc272a3h, 2-amino-3,4-dimethyl benzoic acid
PubChem CID 282450
IUPAC Name 2-amino-3,4-dimethylbenzoic acid
SMILES CC1=CC=C(C(O)=O)C(N)=C1C

Specifications

Melting Point 181°C
Color Yellow
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3,4-dimethylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.