Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3,5-dimethylbenzenesulfonic Acid 97.0+%, TCI America™
SDP

Catalog No. D268025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D268025G 25 g
D2680500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D268025G Supplier TCI America Supplier No. D268025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88-22-2
Molecular Formula C8H11NO3S
Molecular Weight (g/mol) 201.24
MDL Number MFCD00035770
InChI Key CFCXQQUQLZIZPI-UHFFFAOYSA-N
Synonym m-Xylidine-6-sulfonic Acid, 2,4-Dimethylaniline-6-sulfonic Acid
PubChem CID 66608
IUPAC Name 2-amino-3,5-dimethylbenzenesulfonic acid
SMILES CC1=CC(=C(C(=C1)S(=O)(=O)O)N)C

Specifications

Color Yellow
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 201.24
Percent Purity ≥97.0% (HPLC,T)
Chemical Name or Material 2-Amino-3,5-dimethylbenzenesulfonic Acid
EINECSNumber (3)-1984
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.