Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-bromo-5-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. A155525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A155525G 25 g
A15555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A155525G Supplier TCI America Supplier No. A155525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17282-00-7
Molecular Formula C6H7BrN2
Molecular Weight (g/mol) 187.04
MDL Number MFCD00068231
InChI Key NDPKXEWDWTZBDG-UHFFFAOYSA-N
Synonym 2-amino-3-bromo-5-methylpyridine, 2-amino-3-bromo-5-picoline, 6-amino-5-bromo-3-picoline, 3-bromo-5-methyl-2-pyridinamine, 2-amino-5-methyl-3-bromopyridine, 2-pyridinamine, 3-bromo-5-methyl, 3-bromo-5-methyl-pyridin-2-ylamine, 3-bromo-5-methyl-2-pyridylamine, 3-bromo-5-methylpyridin-2-ylamine, 3-bromo-5-methylpyridin-1-ium-2-amine
PubChem CID 519380
IUPAC Name 3-bromo-5-methylpyridin-2-amine
SMILES CC1=CC(=C(N=C1)N)Br

Specifications

Melting Point 72°C
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.