Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-chloro-1,4-naphthoquinone 98.0+%, TCI America™
SDP

Catalog No. A1288500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1288500G 500 g
A128825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1288500G Supplier TCI America Supplier No. A1288500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2797-51-5
Molecular Formula C10H6ClNO2
Molecular Weight (g/mol) 207.61
MDL Number MFCD00001680
InChI Key OBLNWSCLAYSJJR-UHFFFAOYSA-N
Synonym 2-amino-3-chloro-1,4-naphthoquinone, quinoclamine, quinoclamin, mogeton, 06k-quinone, acnq, o 6k-quinone, mogeton granule, mogeton g, 1,4-naphthalenedione, 2-amino-3-chloro
PubChem CID 17748
ChEBI CHEBI:81849
IUPAC Name 2-amino-3-chloro-1,4-dihydronaphthalene-1,4-dione
SMILES NC1=C(Cl)C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 200°C
Quantity 500 g
Formula Weight 207.61
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-chloro-1,4-naphthoquinone
EINECSNumber (4)-0390
RTECSNumber QL7350000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.