Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Amino-3-chlorobenzoic acid, 98%

Click to view available options
Quantity:
25g
100g
6388-47-2
C7H6ClNO2
171.58
MFCD00134229
LWUAMROXVQLJKA-UHFFFAOYSA-N
3-chloroanthranilic acid,benzoic acid, 2-amino-3-chloro,2-amino-3-chloro-benzoic acid,3-chloro-2-aminobenzoic acid,anthranilic acid, 3-chloro,2-amino-3-chlorobenzoicacid,pubchem4639,3-chloroanthranilicacid,acmc-209nj4,amino-3-chlorobenzoic acid
80807
2-amino-3-chlorobenzoic acid
C1=CC(=C(C(=C1)Cl)N)C(=O)O
6388-47-2
C7H6ClNO2
171.58
MFCD00134229
LWUAMROXVQLJKA-UHFFFAOYSA-N
3-chloroanthranilic acid,benzoic acid, 2-amino-3-chloro,2-amino-3-chloro-benzoic acid,3-chloro-2-aminobenzoic acid,anthranilic acid, 3-chloro,2-amino-3-chlorobenzoicacid,pubchem4639,3-chloroanthranilicacid,acmc-209nj4,amino-3-chlorobenzoic acid
80807
2-amino-3-chlorobenzoic acid
C1=CC(=C(C(=C1)Cl)N)C(=O)O
Applications
2-Amino-3-chlorobenzoic acid is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Air & Light Sensitive. Store away from air. Store in the dark. Incompatible with light, air and oxidizing agents.
Sensitivity Air & Light Sensitive
Color Yellow to Brown
Quantity 100g
Melting Point 189°C to 193°C
Beilstein 2090431
Formula Weight 171.58
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Amino-3-chlorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning