Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-3-cyanopyridine 98.0+%, TCI America™
SDP

Catalog No. A244325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A244325G 25 g
A24435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A244325G Supplier TCI America Supplier No. A244325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24517-64-4
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128867
InChI Key YYXDQRRDNPRJFL-UHFFFAOYSA-O
Synonym 2-amino-3-cyanopyridine, 2-aminonicotinonitrile, 2-amino-3-pyridinecarbonitrile, 2-amino-pyridine-3-carbonitrile, 2-amino-nicotinonitrile, 3-pyridinecarbonitrile, 2-amino, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, aminonicotinonitrile, zlchem 506
PubChem CID 818259
IUPAC Name 2-amino-3-cyanopyridin-1-ium
SMILES NC1=[NH+]C=CC=C1C#N

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 119.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-3-cyanopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.