Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4-(2-pyridyl)thiazole, 97%, Thermo Scientific Chemicals

Catalog No. AAH5855409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5855409 10 g
AAH5855404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5855409 Supplier Thermo Scientific Chemicals Supplier No. H5855409

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30235-26-8
Molecular Formula C8H7N3S
Molecular Weight (g/mol) 177.225
MDL Number MFCD00460417
InChI Key BLKHMTAXNXLDJP-UHFFFAOYSA-N
Synonym 4-pyridin-2-yl thiazol-2-amine, 4-pyridin-2-yl-thiazol-2-ylamine, 4-pyridin-2-yl-1,3-thiazol-2-amine, 2-amino-4-2-pyridyl thiazole, 2-thiazolamine, 4-2-pyridinyl, 4-2-pyridinyl-1,3-thiazol-2-amine, 4-2-pyridinyl-2-thiazolamine, 4-2-pyridinyl thiazol-2-amine, 4-2-pyridyl-1,3-thiazole-2-ylamine, 4-pyridin-2-yl-thiazole-2-ylamine
PubChem CID 1092459
IUPAC Name 4-pyridin-2-yl-1,3-thiazol-2-amine
SMILES C1=CC=NC(=C1)C2=CSC(=N2)N

Specifications

Color Brown
Quantity 10 g
Formula Weight 177.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-4-(2-pyridyl)thiazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.