Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile, 97%

Catalog No. AAH3254403
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3254403 1 g
AAH3254409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3254403 Supplier Thermo Scientific Chemicals Supplier No. H3254403
Add to Cart

CAS: 4651-91-6 | C9H10N2S | 178.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4651-91-6
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.253
MDL Number MFCD00128278
InChI Key ADHVMGAFAKSNOM-UHFFFAOYSA-N
Synonym 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitrile, 2-amino-3-cyano-4,5,6,7-tetrahydrobenzo b thiophene, 2-amino-4,5,6,7-tetrahydro-benzo b thiophene-3-carbonitrile, benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydro, 2-amino-4,5,6,7-tetrahydro-1-benzo b thiophene-3-carbonitrile, 2-amino-4,5,6,7-tetrahydrobenzo b thiophene-3-carbonitryle, cambridge id 5130181, aminotetrahydrobenzothiophenecarbonitrile
PubChem CID 78382
IUPAC Name 2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carbonitrile
SMILES C1CCC2=C(C1)C(=C(S2)N)C#N

Specifications

Melting Point 141°C to 144°C
Quantity 1 g
Formula Weight 178.26
Percent Purity 97%
Chemical Name or Material 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-085-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.