Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Amino-4,6-dimethoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. A116525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A116525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A116525G Supplier TCI America Supplier No. A116525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 36315-01-2
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00038832
InChI Key LVFRCHIUUKWBLR-UHFFFAOYSA-N
Synonym 2-amino-4,6-dimethoxypyrimidine, 2-pyrimidinamine, 4,6-dimethoxy, 4,6-dimethoxy-2-pyrimidinamine, 4,6-dimethoxy-2-aminopyrimidine, 4,6-dimethoxy-pyrimidin-2-ylamine, 2-amino-4,6-dimethoxy pyrimidine, 2-amino-4,6-dimethoxy-pyrimidine, 4,6-dimethoxypyrimidine-2-ylamine, maybridge1_002259, zlchem 350
PubChem CID 118946
IUPAC Name 4,6-dimethoxypyrimidin-2-amine
SMILES COC1=CC(=NC(=N1)N)OC

Specifications

Melting Point 98°C
Color White
Quantity 25 g
Formula Weight 155.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-4,6-dimethoxypyrimidine
EINECSNumber (5)-6163
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.